missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard for ICP, ICP-MS 1000 μg/mL (1000 ppm) in 2-5% Nitric Acid (HNO3), Reagecon
BIOPHARMA

Product Code. 11448909 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448909 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448909 Supplier Reagecon Supplier No. PCA2B2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured from ultra-pure metal (99.999%) tested by an ISO17025 Accredited Test Method (INAB Ref: 264T) and raw materials assayed by titration and ICP-MS prior to manufacture.

  • ICP, ICP-MS Reference Standard, ICP, ICP-MS Reference Materia
  • ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
  • ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
  • ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
  • ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
  • ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
  • GHS/OSHA compliant, REACH etc.
  • This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of mass spectroscopy standards), triple distilled acids and triple leached pre-cleaned bottles.
  • The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
  • All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.

Specifications

Description Matrix: 2-5% HNO3
Packaging HDPE Triple-leached
Analyte or Component Names Calcium
Physical Form Liquid
Concentration 1000 µg/mL (1000 ppm)
Technique ICP, ICP-MS, AA, Ion Chromatography, XRF, Cation Measurement
Element Ca
Quantity 250 mL
compliance-icons
Product Identifier
  • CALCIUM ICP STANDARD SOLUTION
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.