missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard For ICP, 1000μg/mL in 2% HCl, SPEX CertiPrep™

Product Code. 10426932 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10426932 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10426932 Supplier SPEX Certiprep Supplier No. PLCA12X
  • ONLINE EXCLUSIVE PRICE
    £155.49
    £125.79 / 500mL
    Save £29.70
    19% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7440-70-2, 7697-37-2, 7732-18-5
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula Ca
MDL Number MFCD00010897 MFCD00085314
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [Ca]
IUPAC Name calcium
Solvent or Matrix 2% HCl
Molecular Weight (g/mol) 40.08
PubChem CID 5460341
Quantity 500 mL
ChEBI CHEBI:29320
Grade Assurance
Chemical Name or Material Calcium Standard
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.