missing translation for 'onlineSavingsMsg'
Learn More

Calcium silicide, tech.

Product Code. 11376978
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11376978 500 g 500g
11366978 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11376978

Brand: Thermo Scientific Alfa Aesar 014676.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12013-56-8 | CaSi2 | 96.248 g/mol

It is an important raw material used in Pharmaceutical intermediates. It is an ideal compound of deoxidizer and desulfurizer which can be used in production of many types of stainless steel. It is also used as inoculants and additive in cast iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12013-56-8
Molecular Formula CaSi2
Molecular Weight (g/mol) 96.248
MDL Number MFCD00015977
InChI Key XXCKHUMTXOCLPT-UHFFFAOYSA-N
Synonym calciumdisilicid, ? 1-silylcalcio-? 1-silanylidyne
PubChem CID 13848147
IUPAC Name calcium;silicon(1-)
SMILES [Si-].[Si-].[Ca+2]

Specifications

Melting Point ∼1000°C
Color Gray
Physical Form Powder
Quantity 500 g
UN Number UN1405
Solubility Information Insoluble in cold water, decomposes in hot water
Formula Weight 96.25
Grade Technical
Concentration or Composition (by Analyte or Components) Ca ≈30%, may contain up to 5% Fe
Sensitivity Moisture sensitive
Density 2.5 g/mL
Chemical Name or Material Calcium silicide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.