missing translation for 'onlineSavingsMsg'
Learn More

Calcium shot, 1cm (0.4 in.) & down, 99% (metals basis), Thermo Scientific™

Catalog Number 11311178
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-70-2
Ca
40.08
MFCD00010897 MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
5460341
CHEBI:29320
calcium
[Ca]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-70-2
Ca
40.08
MFCD00010897 MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
5460341
CHEBI:29320
calcium
[Ca]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
UN Number UN1401
Quantity 100 g
Percent Purity 99%
Odor Odorless
Particle Size 1cm (0.4 in.) & down
Form Shot
Assay (metals basis)
Chemical Name or Material Calcium shot
compliance-icons
Product Identifier
  • Calcium shot
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P233-Keep container tightly closed.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'