Learn More
Calcium-sensing R/CaSR Antibody (HL 1499), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-1830FR
Product Code. 18053955
  • £488.00 / 0.10mL
Description
Calcium-sensing R/CaSR Monoclonal specifically detects Calcium-sensing R/CaSR in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calcium-sensing R/CaSR
Monoclonal
DyLight 680
P41180
CASR
N-terminal region of human Calcium Sensing Receptor. [UniProt# P41180]
Protein G purified
RUO
Primary
Human, Mouse, Rat, Rabbit
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
HL 1499
Western Blot, ELISA
calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441
Mouse
130 kDa
0.1 mL
Cytoskeleton Markers, Extracellular Matrix, GPCR, Growth and Development, Lipid and Metabolism, Neuroscience, Neurotransmission, Signal Transduction
846
Store at 4C in the dark.
IgG1 κ