missing translation for 'onlineSavingsMsg'
Learn More

Calcium Propionate, TRC

Product Code. 30331785
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331785 Supplier Toronto Research Chemicals Supplier No. TRCC1426651G
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propionic acid calcium salt
CAS 4075-81-4
Molecular Formula 2 C3 H5 O2 . Ca
Quantity 1 g
Synonym Propanoic acid, calcium salt (2:1) (ACI), Propanoic acid, calcium salt (9CI), Propionic acid, calcium salt (8CI), Bioban-C, Calcium dipropionate, Calcium propanoate, Calcium propionate, Di(propanoyloxy)calcium, Propionic acid calcium
InChI Formula InChI=1S/2C3H6O2.Ca/c2*1-2-3(4)5;/h2*2H2,1H3,(H,4,5);/q;;+2/p-2
SMILES [Ca+2].CCC(=O)[O-].CCC(=O)[O-]
IUPAC Name calcium;propanoate
Molecular Weight (g/mol) 186.22
Formula Weight 186.0205
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.