missing translation for 'onlineSavingsMsg'
Learn More

Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL

Product Code. 11361419
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361419 50 mL 50mL
11371419 100 mL 100mL
11440360 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11361419 Supplier Thermo Scientific Alfa Aesar Supplier No. 013852.AD
  • ONLINE EXCLUSIVE PRICE
    £41.60
    £37.65 / 50mL
    Save £3.95
    9% Off
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

Calcium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1 , 7732-18-5
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
Solubility Miscible with water
IUPAC Name calcium carbonate
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Calcium carbonate: 0.25%; Water: 94.75%
Chemical Name or Material Calcium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.