missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate Tribasic, MP Biomedicals

Product Code. 11402411 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402411 25 g 25g
11412411 100 g 100g
11422411 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11402411 Supplier MP Biomedicals Supplier No. 0219380525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 12167-74-7
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.17
MDL Number MFCD00010904
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
PubChem CID 14781
IUPAC Name tricalcium diphosphate
SMILES [Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

Quantity 25 g
Formula Weight 502.3
Chemical Name or Material Calcium Phosphate Tribasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.