missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate tribasic, 34-40% Ca

Product Code. 11344257
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 11344257

Brand: Thermo Scientific Alfa Aesar 036731.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12167-74-7 | Ca3O8P2 | 310.17 g/mol

Utilized in powdered spices as an anticaking agent and baby powder. It is the starting material for the production of phosphoric acid and fertilizers. Employed in tissue replacement for repairing bony defects and act as drug carrier for drug delivery to the bone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12167-74-7
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.17
MDL Number MFCD00010904
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
PubChem CID 14781
IUPAC Name tricalcium diphosphate
SMILES [Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 2 kg
Assay Percent Range 34-40% Ca
Solubility Information Partially soluble in water.
Formula Weight 1004.67
Appearance White powder
Chemical Name or Material Calcium phosphate tribasic, tribasic

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.