missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate (pyro), 96% min

Product Code. 11413840
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413840 250 g 250g
11493830 1 kg 1kg
11403840 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413840 Supplier Thermo Scientific Alfa Aesar Supplier No. 089836.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-76-3 | Ca2O7P2 | 254.10 g/mol

Calcium phosphate is used in the production of phosphoric acid and fertilizers. It is used in baking as the acid in a leavening agent. It is also used in cheese products. This compound has also been used in gene transfection of cells. It is also being used in the development of contrast agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-76-3
Molecular Formula Ca2O7P2
Molecular Weight (g/mol) 254.10
MDL Number MFCD00015983
InChI Key JUNWLZAGQLJVLR-UHFFFAOYSA-J
Synonym calcium pyrophosphate, calcium diphosphate, dicalcium diphosphate, unii-x69nu20d19, dicalcium pyrophosphate, alpha-calcium pyrophosphate, calcium phosphate ca2p2o7, diphosphoric acid, calcium salt, calcium pyrophosphate ca2p2o7, diphosphoric acid, calcium salt 1:2
PubChem CID 24632
ChEBI CHEBI:32598
IUPAC Name dicalcium;phosphonato phosphate
SMILES [Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

Melting Point 1230°C
Color White
Physical Form Powder
Quantity 250 g
Solubility Information Soluble in hydrochloric acid and nitric acid. Insoluble in water.
Formula Weight 254.1
Percent Purity ≥96%
Odor Odorless
Density 3.09 g/mL
Chemical Name or Material Calcium phosphate (pyro)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.