missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15597904 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15597904 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15597904 Supplier Avantor J.T.Baker Supplier No. 143005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 7757-93-9
Physical Form Powder
Quantity 2.5 kg
Molecular Formula CaHO4P
MDL Number MFCD00010909
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
SMILES [Ca++].OP([O-])([O-])=O
IUPAC Name calcium;hydrogen phosphate
Molecular Weight (g/mol) 136.06
PubChem CID 24441
ChEBI CHEBI:32596
Formula Weight 136.06
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.30kg
Chemical Name or Material Calcium Phosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.