missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate Monobasic Monohydrate, purum p.a., ≥85% (KT), Honeywell™

Product Code. 15601930 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 15601930

Brand: Honeywell Research Chemicals 210531KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 10031-30-8
Color Colorless
Quantity 1 kg
Molecular Formula CaH6O9P2
Linear Formula Ca(H2PO4)2 · H2O
MDL Number MFCD00149602
UN Number NONH for all modes of transport
Synonym calcium phosphate, monobasic, monohydrate, calcium dihydrogenphosphate hydrate, calcium phosphate monobasic monohydrate, primary calcium phosphate monohydrate, calcium dihydrogenphosphate monohydrate, calcium dihydrogen phosphate monohydrate, calcium bis dihydrogenphosphate monohydrate, monobasic calcium phosphate hydrate jp17, calcium bis dihydrogen phosphate monohydrate, calcium hydrate didihydrogen phosphate
InChI Key ZBZJARSYCHAEND-UHFFFAOYSA-L
SMILES O.OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2]
IUPAC Name calcium;dihydrogen phosphate;hydrate
Molecular Weight (g/mol) 252.065
PubChem CID 10198893
Formula Weight 252.07g/mol
Percent Purity ≥85% (KT)
Grade purum p.a.
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Calcium Phosphate Monobasic Monohydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.