missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate, Honeywell™

Product Code. 15661940 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15661940 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15661940 Supplier Honeywell Research Chemicals Supplier No. 212181KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7758-87-4
Quantity 1 kg
Molecular Formula Ca3O8P2
MDL Number MFCD00015984
UN Number NONH for all modes of transport
Synonym calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]
IUPAC Name tricalcium;diphosphate
Molecular Weight (g/mol) 310.174
PubChem CID 24456
ChEBI CHEBI:9679
Formula Weight 310.18g/mol
Percent Purity ≥96% (calc. as Ca3(PO4)2)
Grade purum p.a.
Chemical Name or Material Calcium Phosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.