missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate dibasic, Puriss. p.a., anhydrous, >98.5% (KT), Solstice

Product Code. 16615763
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16615763 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615763 Supplier Fluka Supplier No. 2117725KG
  • ONLINE EXCLUSIVE PRICE
    £10240.00
    £9436.82 / 25kg
    Save £803.18
    7% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7757-93-9
Color White
CAS Max % 1
Quantity 25 kg
Molecular Formula CaHO4P
Linear Formula CaHPO4
Synonym Calcium hydrogen phosphate
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
SMILES [Ca++].OP([O-])([O-])=O
IUPAC Name calcium hydrogen phosphate
Molecular Weight (g/mol) 136.06
Grade ACS
Chemical Name or Material Calcium phosphate dibasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.