missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate Dibasic, MP Biomedicals

Product Code. 11472401 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472401 100 g 100g
11482401 500 g 500g
11492401 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472401 Supplier MP Biomedicals Supplier No. 0219380480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Quantity 100 g
Formula Weight 136.1
Chemical Name or Material Calcium Phosphate Dibasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.