missing translation for 'onlineSavingsMsg'
Learn More

Calcium perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11331768
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331768 1 kg 1kg
11321768 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331768 Supplier Thermo Scientific Alfa Aesar Supplier No. 011655.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Calcium perchlorate hydrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-36-6
Molecular Formula CaCl2O8
Molecular Weight (g/mol) 238.97
MDL Number MFCD00015971
InChI Key ZQAOXERLHGRFIC-UHFFFAOYSA-L
Synonym calcium perchlorate, calcium diperchlorate, perchloric acid,calcium salt, tetrahydrate 8ci,9ci, hydrated, perchloric acid, calcium salt, perchloric acid, calcium salt 2:1, ca.2clo4, calcium 2+ diperchlorate ion, calcium perchlorate hydrate 250g, perchloric acid calcium salt 2:1e
PubChem CID 61629
IUPAC Name calcium;diperchlorate
SMILES [Ca++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Color White
Physical Form Crystalline
Quantity 1 kg
UN Number UN1455
Formula Weight 238.98 (Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Calcium perchlorate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.