missing translation for 'onlineSavingsMsg'
Learn More

Calcium Oxide, TRC

Product Code. 30342820
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342820 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342820 Supplier Toronto Research Chemicals Supplier No. TRCC14565050G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium oxide
CAS 1305-78-8
Molecular Formula CaO
Quantity 50 g
Synonym Airlock, Bell CML 41, Bell CML(E), Bell CML(P), Borumikku, Burnt lime, CA 602, CAC 20209A, CML 21, CML 31, CML 35, CML 35S, CSQ, Calcia, Calcia (CaO), Calciogreen, Calcium monoxide, Caloxal CPA, Caloxol CP 2, Caloxol W 3, Caltrac
InChI Formula InChI=1S/Ca.O
SMILES O=[Ca]
IUPAC Name oxocalcium
Molecular Weight (g/mol) 56.077
Formula Weight 55.9575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.