missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxide, Reagent Grade

Product Code. 11317618
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317618 500 g 500g
11307618 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317618 Supplier Thermo Scientific Alfa Aesar Supplier No. 033299.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1305-78-8 | CaO | 56.08 g/mol

Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
IUPAC Name oxocalcium
SMILES [O--].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 2572°C
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water with generation of heat,and forming Ca(OH)2,in acids,glycerol. Insoluble in alcoholSoluble in water. Insoluble in methanol,diethyl ether and n-octanol.
Formula Weight 56.08
Grade Reagent
Refractive Index 1.83
Sensitivity Air and moisture sensitive
Density 3.25-3.38
Chemical Name or Material Calcium oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium oxide, Reagent Grade
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Calcium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.