Learn More
Calcium oxide, Puratronic™, 99.998% (metals basis, excluding other alkaline earth and alkali metals 130ppm max), Thermo Scientific Chemicals
Description
Calcium oxide has wide-ranging utility in industry. It is used in making porcelain, cements, mortars, and glass. It is also employed in flue-gas desulfurization in power plants, purifying sugar, drying the solvents/reagents, preparing bleaching powder, calcium carbide, and calcium cyanamide. It also finds use in water softeners, chemical recovery at Kraft pulp mills, biodiesel production, and in petroleum industry for the detection of traces of water. In agriculture it is used to restore the pH of acidic soils (liming). It is useful as a basic catalyst & acid scavenger in several organic transformations, for example, in Arndt-Eistert Synthesis of alpha-diazoketones, synthesis of heterocycles and oximes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 2850°C |
Melting Point | 2572°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.998% (Metal basis) |
Solubility Information | Slightly soluble in water. Insoluble in ethanol. Soluble in glycerol. |
IUPAC Name | oxocalcium |
Formula Weight | 56.08 |
Odor | Odorless |
Show More |
RUO – Research Use Only