missing translation for 'onlineSavingsMsg'
Learn More

Calcium Oxide, Extra Pure, SLR, Powder, Fisher Chemical

Product Code. 10305030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10305030 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10305030 Supplier Fisher Chemical Supplier No. C/2120/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1kg, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2850°C
Melting Point 2570°C
CAS 1305-78-8
Color Beige
pH 12.5
Physical Form Powder/Solid
Quantity 1 kg
Molecular Formula CaO
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES [O--].[Ca++]
IUPAC Name calcium oxidandiide
Molecular Weight (g/mol) 56.08
Formula Weight 56.08g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium oxide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301-IF SWALLOWED:.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.