missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxide, 99.95% (metals basis)

Product Code. 11410041
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410041 50 g 50g
11420041 250 g 250g
11400041 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410041 Supplier Thermo Scientific Alfa Aesar Supplier No. 044776.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1305-78-8 | CaO | 56.08 g/mol

Calcium oxide has wide-ranging utility in industry. It is used in making porcelain, cements, mortars and glass. It is also employed in flue-gas desulfurization in power plants, purifying sugar, drying the solvents/reagents, preparing bleaching powder, calcium carbide and calcium cyanamide. It also finds use in water softeners, chemical recovery at Kraft pulp mills, biodiesel production and in the petroleum industry for the detection of traces of water. In agriculture, it is used to restore the pH of acidic soils (liming). It is useful as a basic catalyst and an acid scavenger in several organic transformations, for example, in Arndt-Eistert synthesis of alpha-diazoketones, synthesis of heterocycles and oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
SMILES [O--].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 2572°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.95% (Metal basis)
Solubility Information Soluble in water. Insoluble in methanol,diethyl ether and n-octanol.
IUPAC Name oxocalcium
Formula Weight 56.08
Refractive Index 1.83
Sensitivity Air and moisture sensitive
Appearance White powder
Density 3.25-3.37 g/cm3 at 20°C
Chemical Name or Material Calcium oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.