missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxide, 97+%, for analysis, powder

Product Code. 11924161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11924161 2.5 kg Plastic bottle 2.5kg
10381351 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11924161 Supplier Thermo Scientific Acros Supplier No. 422830025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calcium oxide, 97+%, CaO, CAS Number-1305-78-8, gebrannter kalk, calxyl, calcia, burnt lime, calcium oxide, airlock, calcium monoxide, calx, unslaked lime, 1kg, 2.0% max., 97% min. (CaO) on dry substance (Complexometry), 2850 deg.C, CHEBI:31344, White or Yellow, 215-138-9, 56.08, Analysis | CAS: 1305-78-8 | CaO | 56.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
SMILES [O--].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 2570°C
Color White or Yellow
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 97% min. (CaO) on dry substance (Complexometry)
Linear Formula CaO
Merck Index 15,1688
Solubility Information Solubility in water: reacts. Other solubilities: soluble in acids (reacts),soluble in glycerol,insoluble in alcohol
IUPAC Name oxocalcium
Formula Weight 56.08
Percent Purity ≥97%
Grade Analytical
Loss on Ignition 5% max.
Packaging Plastic bottle
Chemical Name or Material Calcium oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Calcium oxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: calcium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.