missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxalate monohydrate, Puratronic™, 99.9985% (metals basis), (excluding 25ppm max alkali earths)

Product Code. 11332108
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332108 25 g 25g
11322108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332108 Supplier Thermo Scientific Alfa Aesar Supplier No. 013007.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5794-28-5 | C2H2CaO5 | 146.11 g/mol

Calcium oxalate monohydrate is used as a precursor in the production of oxalic acid, organic oxalates, ceramic glazes, and for rare-earth-metal separation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-28-5
Molecular Formula C2H2CaO5
Molecular Weight (g/mol) 146.11
MDL Number MFCD00150039
InChI Key LQHWSGSWNOHVHO-UHFFFAOYSA-L
Synonym calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:?
PubChem CID 165350
IUPAC Name calcium;oxalate;hydrate
SMILES O.[Ca++].[O-]C(=O)C([O-])=O

Specifications

Melting Point 200°C
Color White
Physical Form Granules, Powder
Quantity 25 g
Assay Percent Range 99.9985% (Metal basis)
Solubility Information Slightly soluble in water.
Formula Weight 146.11 (128.10 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 2.12
Chemical Name or Material Calcium oxalate monohydrate, H2O at 200°
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.