missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Nitrate Tetrahydrate, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Calcium Nitrate Tetrahydrate |
| CAS | 13477-34-4 |
| Molecular Formula | Ca(NO3)2 4H2O |
| Quantity | 100 g |
| Synonym | Calcium Dinitrate Tetrahydrate, Calcium Nitrate (Ca(NO3)2) Tetrahydrate, Calcium Nitrate Tetrahydrate, Nitric Acid Calcium Salt (2:1) Tetrahydrate |
| InChI Formula | InChI=1S/Ca.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;; |
| SMILES | O.O.O.O.[Ca+2].[O-][N+](=O)[O-].[O-][N+](=O)[O-] |
| IUPAC Name | calcium;dinitrate;tetrahydrate |
| Molecular Weight (g/mol) | 236.15 |
| Formula Weight | 235.9805 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction