missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Nitrate Tetrahydrate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Unit Size:
1kg
250g
500g
5kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 1 kg |
| Linear Formula | Ca(NO3)2 · 4H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 236.15g/mol |
| Percent Purity | 99 to 103% |
| Grade | ACS/puriss. p.a. |
| Chemical Name or Material | Calcium Nitrate Tetrahydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction