missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate tetrahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Product Code. 11361749
Click to view available options
Quantity:
500 g
2 kg

Manufacturing of other nitrates, fertilizers, matches

Calcium nitrate tetrahydrate is used as nitrogenous fertilizer and corrosive inhibitor in diesel fuels. It is utilized in plant cell for culture experiments. It is also used in cooling baths and waste water handling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.146
MDL Number MFCD00149604
InChI Key ICSSIKVYVJQJND-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium;dinitrate;tetrahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]

Specifications

Boiling Point 132°C (decomposition)
Melting Point 42°C
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0-103.0%
UN Number UN1454
Merck Index 14,1682
Solubility Information Soluble in water,alcohol,acetone and ammonia.
Formula Weight 236.15 (164.09 Anhydrous)
Percent Purity 99.0 to 103.0%
Grade ACS
Sensitivity Hygroscopic
Density 1.82
Chemical Name or Material Calcium nitrate, tetrahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'