missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, 99.0 to 103.0%, ACS Reagent, Honeywell Fluka™

Product Code. 15622360 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15622360

Brand: Honeywell-Fluka 237124500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
MDL Number MFCD00149604
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Melting Point 44°C
pH 4 to 7 at 20°C
Quantity 500 g
Linear Formula Ca(NO3)2 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 236.15g/mol
Percent Purity 99.0 to 103.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.86g/mL at 25°C
Chemical Name or Material Calcium Nitrate Tetrahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.