missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate hydrate, Puratronic™, 99.995% (metals basis)

Product Code. 11340929
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340929 5 g 5g
11320929 25 g 25g
11330929 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11340929 Supplier Thermo Scientific Alfa Aesar Supplier No. 010683.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 35054-52-5 | CaN2O6 | 164.09 g/mol

Calcium nitrate hydrate is used as a nitrate enriching salt. It is also used as a component in fertilizers, waste water treatment to prevent odor and inset accelerating concrete admixtures. Further, it is used for regenerable cold packs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35054-52-5
Molecular Formula CaN2O6
Molecular Weight (g/mol) 164.09
MDL Number MFCD00149604
InChI Key ZCCIPPOKBCJFDN-UHFFFAOYSA-N
Synonym calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci
PubChem CID 16211464
SMILES [Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 132°C (decomposition)
Melting Point 42°C
Physical Form Crystalline Aggregates
Quantity 5 g
UN Number UN1454
Merck Index 14,1682
IUPAC Name calcium;dinitrate;hydrate
Formula Weight 164.09 (Anhydrous)
Percent Purity 99.995%
Grade Puratronic
Sensitivity Hygroscopic
Density 1.82g/mL
Assay (metals basis)
Chemical Name or Material Calcium nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.