missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Calcium nitrate hydrate is used as a nitrate enriching salt. It is also used as a component in fertilizers, waste water treatment to prevent odor and inset accelerating concrete admixtures. Further, it is used for regenerable cold packs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 132°C (decomposition) |
Melting Point | 42°C |
Physical Form | Crystalline Aggregates |
Quantity | 100 g |
UN Number | UN1454 |
Merck Index | 14,1682 |
Solubility Information | Soluble in water. |
IUPAC Name | calcium dinitrate |
Formula Weight | 164.09 (Anhydrous) |
Percent Purity | 99.995% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'