missing translation for 'onlineSavingsMsg'
Learn More

Calcium molybdenum oxide, 99%

Product Code. 11330278
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11330278

Brand: Thermo Scientific Alfa Aesar 043686.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-82-4 | CaMoO4 | 200.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-82-4
Molecular Formula CaMoO4
Molecular Weight (g/mol) 200.02
MDL Number MFCD00015976
InChI Key BIOOACNPATUQFW-UHFFFAOYSA-N
Synonym calcium molybdate, molybdate, calcium, calcium molybdate vi, unii-q03c83v2an, calcium molybdenum oxide, molybdic acid h2moo4 , calcium salt 1:1, molybdate moo42-, calcium 1:1 , t-4, molybdate moo42-, calcium 1:1 , beta-4, calcium dioxido dioxo molybdenum, calcium 2+ molybdate
PubChem CID 24620
SMILES [Ca++].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 965°C
Color White
Physical Form Powder
Quantity 100 g
Merck Index 14,1681
IUPAC Name calcium dioxomolybdenumbis(olate)
Formula Weight 200.01
Percent Purity 99%
Odor Odorless
Density 4.45 g/mL
Chemical Name or Material Calcium molybdenum oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.