missing translation for 'onlineSavingsMsg'
Learn More

Calcium Methoxide, TRC

Product Code. 30339975
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30339975 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339975 Supplier Toronto Research Chemicals Supplier No. TRCC145656500MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium Methoxide
CAS 2556-53-8
Molecular Formula CH4O • 1/2(Ca)
Quantity 500 mg
Synonym AKC 165, Calcium Dimethoxide
InChI Formula InChI=1S/CH4O.Ca/c1-2;/h2H,1H3;
SMILES [Ca].OC
IUPAC Name methanol, calcium salt
Molecular Weight (g/mol) 52.01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.