missing translation for 'onlineSavingsMsg'
Learn More

Calcium metaborate, 99.992% (metals basis)

Product Code. p-7049132
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11322088

Brand: Thermo Scientific Alfa Aesar 012988.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13701-64-9 | B2CaO4 | 125.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13701-64-9
Molecular Formula B2CaO4
Molecular Weight (g/mol) 125.694
MDL Number MFCD00049397
InChI Key ILOKQJWLMPPMQU-UHFFFAOYSA-N
Synonym calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
PubChem CID 10290800
IUPAC Name calcium;oxido(oxo)borane
SMILES B(=O)[O-].B(=O)[O-].[Ca+2]

Specifications

Color White
Physical Form Powder
Quantity 5 g
Linear Formula Ca(BO2)2
Formula Weight 125.7
Percent Purity 99.992%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Calcium metaborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.