missing translation for 'onlineSavingsMsg'
Learn More

Calcium metaborate, 99.992% (metals basis)

Product Code. 11322088
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322088 5 g 5g
11332088 25 g 25g
11342088 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11322088 Supplier Thermo Scientific Alfa Aesar Supplier No. 012988.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13701-64-9 | B2CaO4 | 125.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13701-64-9
Molecular Formula B2CaO4
Molecular Weight (g/mol) 125.694
MDL Number MFCD00049397
InChI Key ILOKQJWLMPPMQU-UHFFFAOYSA-N
Synonym calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
PubChem CID 10290800
IUPAC Name calcium;oxido(oxo)borane
SMILES B(=O)[O-].B(=O)[O-].[Ca+2]

Specifications

Color White
Physical Form Powder
Quantity 5 g
Linear Formula Ca(BO2)2
Formula Weight 125.7
Percent Purity 99.992%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Calcium metaborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.