missing translation for 'onlineSavingsMsg'
Learn More

Calcium metaborate, 99.992% (metals basis)

Product Code. 11332088
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Conditionnement:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11332088 25 g 25g
11322088 5 g 5g
11342088 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 11332088 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur 012988.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

CAS: 13701-64-9 | B2CaO4 | 125.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 13701-64-9
Molecular Formula B2CaO4
Molecular Weight (g/mol) 125.694
MDL Number MFCD00049397
InChI Key ILOKQJWLMPPMQU-UHFFFAOYSA-N
Synonym calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
PubChem CID 10290800
IUPAC Name calcium;oxido(oxo)borane
SMILES B(=O)[O-].B(=O)[O-].[Ca+2]

Spécification

Color White
Physical Form Powder
Quantity 25 g
Linear Formula Ca(BO2)2
Formula Weight 125.7
Percent Purity 99.992%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Calcium metaborate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.