missing translation for 'onlineSavingsMsg'
Learn More

Calcium Ionophore A23187 (Free Acid), Fisher BioReagents

Product Code. 10284963
Change view
Click to view available options
Poly Micro Tube; 1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10284963 Poly Micro Tube; 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10284963 Supplier Fisher BioReagents Supplier No. BP5951
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Isolated from Streptomyces chartreusensis

This widely used ionophore increases the permeability of biological membranes for bivalent cations.
TRUSTED_SUSTAINABILITY

Specifications

CAS 52665-69-7
Melting Point 182°C
Color White
Packaging Poly Micro Tube
Molecular Formula C29H37N3O6
MDL Number MFCD00151202,MFCD00151202
Quantity 1 mg
Merck Index 15, 1642
Specific Rotation -50 to -56° (+ or -)
Synonym calcimycin, antibiotic a23187, ionophore a23187, calcium ionophore a23187, 4-benzoxazolecarboxylic acid, 5-methylamino-2-3,9,11-trimethyl-8-1-methyl-2-oxo-2-1h-pyrrol-2-yl ethyl-1,7-dioxaspiro 5.5 undec-2-yl methyl-, 6s-6alpha 2s*,3s* ,8beta r* ,9beta,11alpha, spectrum_001976, upcmld-dp093, upcmld-dp093:001, upcmld-dp093:002, upcmld-dp093:003
InChI Key HIYAVKIYRIFSCZ-CVXKHCKVSA-N
SMILES CNC1=CC=C2OC(C[C@@H]3OC4(CC[C@@H]3C)O[C@H](C(C)C(=O)C3=CC=CN3)[C@H](C)C[C@H]4C)=NC2=C1C(O)=O
IUPAC Name 5-(methylamino)-2-{[(2S,3S,8S,9R,11R)-3,9,11-trimethyl-8-[1-oxo-1-(1H-pyrrol-2-yl)propan-2-yl]-1,7-dioxaspiro[5.5]undecan-2-yl]methyl}-1,3-benzoxazole-4-carboxylic acid
Molecular Weight (g/mol) 523.63
PubChem CID 97043288
Formula Weight 523.6
Percent Purity 98% min.
Physical Form Solid
Chemical Name or Material Calcium Ionophore A23187
Show More Show Less
compliance-icons
Product Identifier
  • Calcium ionophore A23187
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.