missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11359788
Click to view available options
Quantity:
25 g
100 g
10102-68-8
CaI2
293.887
MFCD00010910
UNMYWSMUMWPJLR-UHFFFAOYSA-L
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
5462722
CHEBI:81718
calcium;diiodide
[Ca+2].[I-].[I-]
10102-68-8
CaI2
293.887
MFCD00010910
UNMYWSMUMWPJLR-UHFFFAOYSA-L
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
5462722
CHEBI:81718
calcium;diiodide
[Ca+2].[I-].[I-]

Used in chemical research and as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1100°C
Melting Point 784°C
Color White to Yellow
Physical Form Beads
Quantity 25 g
Merck Index 14,1677
Solubility Information Soluble in water,ethyl alcohol,methyl alcohol and acetone. Insoluble in ethyl ether.
Formula Weight 293.89
Percent Purity 99.999%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Density 3.956 g/mL
Assay (metals basis)
Chemical Name or Material Calcium iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'