missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide, ultra dry, 99.99% (metals basis)

Product Code. 11325907
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325907 500 g 500g
11315907 25 g 25g
11335907 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11325907 Supplier Thermo Scientific Alfa Aesar Supplier No. 043674.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Light Sensitive and Hygroscopic | CAS: 10102-68-8 | CaI2 | 293.887 g/mol

Calcium iodide is used in photography. It is also used in cat food as a source of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-68-8
Molecular Formula CaI2
Molecular Weight (g/mol) 293.887
MDL Number MFCD00010910
InChI Key UNMYWSMUMWPJLR-UHFFFAOYSA-L
Synonym calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
PubChem CID 5462722
ChEBI CHEBI:81718
IUPAC Name calcium;diiodide
SMILES [Ca+2].[I-].[I-]

Specifications

Boiling Point 1100°C
Melting Point 784°C
Color White to Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,methanol,ethanol and acetone. Insoluble in ether.
Formula Weight 293.89
Sensitivity Hygroscopic; Light sensitive
Appearance Off-white to pale yellow powder or beads
Density 3.956
Chemical Name or Material Calcium iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.