missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide hydrate

Product Code. 11309287
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11309287

Brand: Thermo Scientific Alfa Aesar 040136.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71626-98-7 | CaI2 | 293.89 g/mol

Calcium iodide finds use in the preparation of complex used in polymerization and hydroamination as well as serving as a matrix for luminescent materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71626-98-7
Molecular Formula CaI2
Molecular Weight (g/mol) 293.89
MDL Number MFCD00149622
InChI Key UNMYWSMUMWPJLR-UHFFFAOYSA-L
IUPAC Name calcium diiodide
SMILES [Ca++].[I-].[I-]

Specifications

Melting Point 42°C (decomposition)
Color White to Gray
Physical Form Lump
Quantity 500 g
Merck Index 14,1677
Solubility Information Freely soluble in water and alcohol
Formula Weight 293.89 (Anhydrous)
Sensitivity Hygroscopic
Density 2.55 g/mL
Chemical Name or Material Calcium iodide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.