missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide hydrate, Puratronic™, 99.9965% (metals basis)

Product Code. 11356138
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356138 25 g 25g
11366138 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356138 Supplier Thermo Scientific Alfa Aesar Supplier No. 011117.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71626-98-7 | CaI2 | 293.89 g/mol

Calcium iodide hydrate used in photography. It is also used in cat food as a source of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71626-98-7
Molecular Formula CaI2
Molecular Weight (g/mol) 293.89
MDL Number MFCD00149622
InChI Key UNMYWSMUMWPJLR-UHFFFAOYSA-L
IUPAC Name calcium diiodide
SMILES [Ca++].[I-].[I-]

Specifications

Melting Point 42°C (decomposition)
Color Yellow
Physical Form Crystalline
Quantity 25 g
Merck Index 14,1677
Solubility Information Freely soluble in water and alcohol
Formula Weight 293.89 (Anhydrous)
Percent Purity 99.9965%
Grade Puratronic
Sensitivity Hygroscopic
Density 2.55 g/mL
Assay (metals basis)
Chemical Name or Material Calcium iodide hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.