Learn More
Calcium iodide, anhydrous, 99.5% (metals basis)
Descrizione
Calcium iodide, anhydrous is used as a drug in upper respiratory combinations, expectorants. And it is also used as Laboratory chemicals, Manufacture of substances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Boiling Point | 1100°C |
| Melting Point | 784°C |
| Physical Form | Powder |
| Quantity | 100 g |
| Sensitivity | Hygroscopic; Light sensitive |
| Assay Percent Range | 99.5% (Metals basis) |
| Density | 3.956 |
| Merck Index | 14,1677 |
| Solubility Information | Freely soluble in water,methanol,ethanol,acetone; Insoluble in ether |
| Formula Weight | 293.89 |
| Vedi altri risultati |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.