missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hypochlorite, Technical grade, Honeywell™ Fluka™

Product Code. p-7120283 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 15691930

Brand: Honeywell-Fluka 2113895G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-54-3
Molecular Formula CaCl2O2
Molecular Weight (g/mol) 142.976
MDL Number MFCD00010900
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
Synonym calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor
PubChem CID 24504
IUPAC Name calcium;dihypochlorite
SMILES [O-]Cl.[O-]Cl.[Ca+2]

Specifications

Melting Point 100°C
Color Beige
Quantity 5 g
Linear Formula Ca(OCl)2
UN Number UN1748
Formula Weight 142.98g/mol
Grade Technical
Packaging Poly Bottle
Appearance Powder
Density 2.35g/mL at 25°C
Chemical Name or Material Calcium Hypochlorite
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.