missing translation for 'onlineSavingsMsg'
Learn More

Calcium hypochlorite, tech.

Product Code. 15431817
Change view
Click to view available options
Quantity:
10 g
250 g
1 kg
Unit Size:
1000g
10g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431817 1 kg 1000g
11319148 10 g 10g
11339148 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431817 Supplier Thermo Scientific Alfa Aesar Supplier No. 042548.0B
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Use in Deodorant, Oxidizing Agent, Bleaching Agent | CAS: 7778-54-3 | CaCl2O2 | 142.976 g/mol

Deodorant, oxidizing agent, bleaching agentCalcium hypochlorite is used for swimming pool sanitation and other water treatment plants. It is also used to disinfect drinking water, equipments in the industry and surfaces and equipments in kitchens. Further, it is used to bleach textiles and paper. It is also employed in bathroom cleaners and laundry detergents. It is used as an oxidizing agent in organic chemistry. It is involved in the cleavage of glycols, alfa-hydroxy carboxylic acids and keto acids to yield fragmented aldehydes or carboxylic acids respectively. It plays an important role in the haloform reaction to prepare chloroform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-54-3
Molecular Formula CaCl2O2
MDL Number MFCD00010900
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
Synonym calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor
PubChem CID 24504

Specifications

Melting Point 100°C
Color White
Physical Form Granules
Quantity 1 kg
Appearance White granular powder
Density 2.35 g/cm3 (at 20°C)
Solubility Information Soluble aq. soln..
SMILES [Ca++].[O-]Cl.[O-]Cl
IUPAC Name calcium dihypochlorite
Molecular Weight (g/mol) 142.98
Formula Weight 142.98
Chemical Name or Material Calcium hypochlorite
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Calcium hypochlorite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Calcium hypochlorite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.