missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, TRC

Product Code. 30346620
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346620 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346620 Supplier Toronto Research Chemicals Supplier No. TRCC145580100G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium hydroxide
CAS 1305-62-0
Molecular Formula Ca H2 O2
Quantity 100 g
Synonym Calcium hydroxide, Calcium hydroxide (8CI), A-Rock, BTTE, Biocalc, CH 2N, CLS-B, Cal-Z, Calbit, Calbreed, Calcicure, Calcium Hydroxide Plus Points, Calcium dihydroxide, Caldic 1000, Caldic 2000, Calen, Calvit, Carboxide, Ecolomaster, HA 20, HA 20 (hydroxide), HP-XL, High Purity Calcium Hydroxide CH, Hydralime, Hydrated Lime, JIS-R 9001, Kalic, Kalkhydrate, Karutekku LT, Kentoku K 100, Limbux, Lime S, Lime Type S, Lime Hydrate, Lime Milk, Lime water, LoFloSorb, Microstar T, Milk of Lime, NICC 3000, NICC 5000, Nekablanc 0, Nekapur 2, Omix C, Precal 54, Rhein CF 1, Rhenofit CF, SA 074, SA 149, Slaked Lime, Soprocal, Sorbacal SP, Sorbacal SPS, Super Microstar, Superia, TP 2B, Tamakalk-Spongiacal, TempCanal, Ultradent-UltraCal XS, Vitacal H, Yukijirushi Shosekkai, Yukijirushisakanyo
InChI Formula InChI=1S/Ca.2H2O/h;2*1H2/q+2;;/p-2
SMILES O[Ca]O
IUPAC Name dihydroxycalcium
Molecular Weight (g/mol) 74.09
Formula Weight 73.9681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.