missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hydroxide, TRC
Specifications
Specifications
| Chemical Name or Material | Calcium hydroxide |
| CAS | 1305-62-0 |
| Molecular Formula | Ca H2 O2 |
| Quantity | 5 g |
| Synonym | Calcium hydroxide, Calcium hydroxide (8CI), A-Rock, BTTE, Biocalc, CH 2N, CLS-B, Cal-Z, Calbit, Calbreed, Calcicure, Calcium Hydroxide Plus Points, Calcium dihydroxide, Caldic 1000, Caldic 2000, Calen, Calvit, Carboxide, Ecolomaster, HA 20, HA 20 (hydroxide), HP-XL, High Purity Calcium Hydroxide CH, Hydralime, Hydrated Lime, JIS-R 9001, Kalic, Kalkhydrate, Karutekku LT, Kentoku K 100, Limbux, Lime S, Lime Type S, Lime Hydrate, Lime Milk, Lime water, LoFloSorb, Microstar T, Milk of Lime, NICC 3000, NICC 5000, Nekablanc 0, Nekapur 2, Omix C, Precal 54, Rhein CF 1, Rhenofit CF, SA 074, SA 149, Slaked Lime, Soprocal, Sorbacal SP, Sorbacal SPS, Super Microstar, Superia, TP 2B, Tamakalk-Spongiacal, TempCanal, Ultradent-UltraCal XS, Vitacal H, Yukijirushi Shosekkai, Yukijirushisakanyo |
| InChI Formula | InChI=1S/Ca.2H2O/h;2*1H2/q+2;;/p-2 |
| SMILES | O[Ca]O |
| IUPAC Name | dihydroxycalcium |
| Molecular Weight (g/mol) | 74.09 |
| Formula Weight | 73.9681 |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?