missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydroxide, Purum p.a., ≥96.0% (T), Solstice

Product Code. 16625763
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16625763 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16625763 Supplier Fluka Supplier No. 2118125KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 580°C
CAS 1305-62-0
Color Colorless
CAS Max % ≤ 100%
pH 12 to 13
Quantity 25 kg
Molecular Formula CaH2O2
Linear Formula Ca(OH)2
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES [OH-].[OH-].[Ca++]
IUPAC Name calcium dihydroxide
Molecular Weight (g/mol) 74.09
Grade Reagent
Density 2.24 g/cm3 (20°C)
Chemical Name or Material Calcium hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.