missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15547904 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2 kg
Packaging:
Poly Pail
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15547904 2 kg Poly Pail 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15547904 Supplier Avantor J.T.Baker Supplier No. 137205

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 1305-62-0
Physical Form Powder
Quantity 2 kg
Molecular Formula CaH2O2
MDL Number MFCD00010901
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES [OH-].[OH-].[Ca++]
IUPAC Name calcium;dihydroxide
Molecular Weight (g/mol) 74.09
PubChem CID 6093208
ChEBI CHEBI:31341
Formula Weight 74.09
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Pail
Specific Gravity 1L = 2.24kg
Chemical Name or Material Calcium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.