missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 450°C |
| Color | White |
| Quantity | 100 g |
| Linear Formula | Ca(OH)2 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 74.09g/mol |
| Percent Purity | ≥95.0% |
| Grade | ACS Reagent |
| Packaging | Poly Bottle |
| Appearance | Powder |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?