missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, ≥95.0%, ACS Reagent, Honeywell Fluka™

Product Code. 15632430 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15632430 500 g Poly Bottle 500g
15612430 100 g Poly Bottle 100g
15622430 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15632430 Supplier Honeywell-Fluka Supplier No. 239232500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥95.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium;dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Melting Point 450°C
Color White
Quantity 500 g
Linear Formula Ca(OH)2
UN Number NONH for all modes of transport
Formula Weight 74.09g/mol
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 2.24g/mL at 25°C
Chemical Name or Material Calcium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.