missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydrogen phosphate dihydrate, 98% min

Product Code. 11329397
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329397 5 kg 5kg
11319397 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329397 Supplier Thermo Scientific Alfa Aesar Supplier No. 040233.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-77-7 | CaH5O6P | 172.086 g/mol

Calcium hydrogen phosphate dihydrate is used in the manufacture of pharmaceutical tablets and as a source of calcium and phosphorus in nutritional supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-77-7
Molecular Formula CaH5O6P
Molecular Weight (g/mol) 172.086
MDL Number MFCD00149621
InChI Key XAAHAAMILDNBPS-UHFFFAOYSA-L
Synonym calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen
PubChem CID 104805
ChEBI CHEBI:4496
IUPAC Name calcium;hydrogen phosphate;dihydrate
SMILES O.O.OP(=O)([O-])[O-].[Ca+2]

Specifications

Melting Point 109°C (in -H2O)
Color White
Physical Form Powder
Quantity 5 kg
Linear Formula CaHPO4·2H2O
Merck Index 14,1692
Solubility Information Slightly soluble in water; Soluble in dilute hydrochloric, nitric, and acetic acid; Insoluble in alcohol
Formula Weight 172.09 (136.06 Anhydrous)
Percent Purity ≤98%
Odor Odorless
Density 2.306 g/mL
Chemical Name or Material Calcium hydrogen phosphate dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.