missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydride, ca. 93%, extra pure, approx. 1-20 mm grain size

Product Code. 10246350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246350 500 g Plastic bottle 500g
10752431 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10246350 Supplier Thermo Scientific Acros Supplier No. 196795000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7789-78-8 | CaH2 | 42.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-78-8
Molecular Formula CaH2
Molecular Weight (g/mol) 42.09
MDL Number MFCD00010897
InChI Key FAQLAUHZSGTTLN-UHFFFAOYSA-N
IUPAC Name calcium dihydride
SMILES [CaH2]

Specifications

Melting Point 675.0°C
Color Gray
Physical Form Granular Powder
Quantity 500 g
Assay Percent Range 92% min. (Total Ca)
Linear Formula CaH2
UN Number 1404
Fieser 01,105
Merck Index 15, 1674
Solubility Information Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts
Formula Weight 42.09
Percent Purity ∼93%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 1.9
Density 1.9000g/mL
Chemical Name or Material Calcium hydride, ca. 93%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.